Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Short Visor for X5000 Safety Helmet

Catalog No. 19071093 Shop All 3M Products
Change view
Click to view available options
Color:
Amber
Clear
Gray
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-071-093 Gray
19-071-094 Amber
19-071-122 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-071-093 Supplier 3M™ Supplier No. 7100176970
Add to Cart
Add to Cart

Short Visor is molded out of polycarbonate for impact protection in a variety of applications.

  • Features an anti-fog, anti-scratch coating.
  • Available in clear, grey, and amber.
  • Fits 3M™ SecureFit™ X5000 Series Safety Helmets
  • 3M Catalog# X5-SV01 , 3M Catalog# X5-SV02 , 3M Catalog# X5-SV03

TRUSTED_SUSTAINABILITY

Specifications

Color Gray
For Use With (Application) Demolition, Forestry, Grinding, Remediation
Material Polycarbonate
Model X5-SV02
Product Type Short Visor
Quantity 10/Cs.
Size One Size Fits Most
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.