Learn More
Cayman Chemical Short-chain Fatty Acid MaxSpec LC-MS Mixture, 1 ea
SDP
Catalog No. 502966210
Encompass_Preferred
Contains various short-chain fatty acids (≤12:0); designed for direct use in LC-MS applications
Catalog No. 50-296-6210 Supplier Cayman Chemical Supplier No. 188241 EA
Contains various short-chain fatty acids (≤12:0); designed for direct use in LC-MS applications