Learn More
Cayman Chemical ShortchaIn Faty ACD MIur 100mL
SDP
Catalog No. 502941966
Encompass_Preferred
A mixture of ten short-chain and volatile fatty acids; intended for use as a reference standard for the detection of short-chain fatty acids in various sample types by GC-MS
Catalog No. 50-294-1966 Supplier Cayman Chemical Supplier No. 28682100 ML
A mixture of ten short-chain and volatile fatty acids; intended for use as a reference standard for the detection of short-chain fatty acids in various sample types by GC-MS