Learn More
New England Biolabs, Inc. ShortCut RNase III – 200 units
SDP
Catalog No. 50811661
Encompass
ShortCut RNase III, used with its manganese-containing reaction buffer, converts long double-stranded RNA into a heterogeneous mix of short (18-25 bp) interfering RNAs (siRNA) suitable for RNA interference in mammalian cells. 1.5 units (1 µl) of ShortCut RNase III is sufficient to convert 1µ g of dsRNA into siRNA suitable for RNA interference in mammalian cells.
Catalog No. 50-811-661 Supplier New England Biolabs, Inc. Supplier No. M0245S
ShortCut RNase III, used with its manganese-containing reaction buffer, converts long double-stranded RNA into a heterogeneous mix of short (18-25 bp) interfering RNAs (siRNA) suitable for RNA interference in mammalian cells. 1.5 units (1 µl) of ShortCut RNase III is sufficient to convert 1µ g of dsRNA into siRNA suitable for RNA interference in mammalian cells.