Learn More
Braintree Scientific Shorti Tail Vein Injection Tube, Mouse, Red Tube, 1.25"""" ID
SDP
Catalog No. 501955892
Encompass
NEW! Red Short Base (Shorti) Tail Vein Injection Tube Tail vein restraint system,mounted on a widerbaseplate allowing the tail to drop vertically for greater range of motion for venipuncture. Use when intending to mount the tube above (e.g. on blocks) or at the edge of a work station/table.Advantages of the red tube vs. the opaque tube Laboratory rodents are largely color blind, so a red tube simply looks like a comforting shelter. The Red Tube is transparent, allowing the investigator to observe the animal inside for normal body position and breathing. Improved comfort for mice during acute experimental procedures without sacrificing safety or welfare. **Sold for research purposes only and not for human use**
Catalog No. 50-195-5892 Supplier Braintree Scientific Supplier No. SHORTISTD
 Restricted Item
Add to Cart
Add to Cart
NEW! Red Short Base (Shorti) Tail Vein Injection Tube Tail vein restraint system,mounted on a widerbaseplate allowing the tail to drop vertically for greater range of motion for venipuncture. Use when intending to mount the tube above (e.g. on blocks) or at the edge of a work station/table.Advantages of the red tube vs. the opaque tube Laboratory rodents are largely color blind, so a red tube simply looks like a comforting shelter. The Red Tube is transparent, allowing the investigator to observe the animal inside for normal body position and breathing. Improved comfort for mice during acute experimental procedures without sacrificing safety or welfare. **Sold for research purposes only and not for human use**