Learn More
Bellco Glass, Inc. Shortie Pipet,25x.5mL Cotton Plug Top
SDP
Catalog No. 501944638
Encompass_Preferred
Bellco manufactures all precision pipettes from type 1 class a Borosilicate glass. All bellco pipettes feature tempered tips and permanent silk screened graduations to withstand repeated usage and alkali washings. Shorter length facilitates use under biological hood. MADE IN THE USA Case qty-18 TOLERANCE +-(ML):0.25OAL (MM):230COLOR CODE: White Graduation: 0.5
Catalog No. 50-194-4638 Supplier Bellco Glass, Inc. Supplier No. 122625050
Bellco manufactures all precision pipettes from type 1 class a Borosilicate glass. All bellco pipettes feature tempered tips and permanent silk screened graduations to withstand repeated usage and alkali washings. Shorter length facilitates use under biological hood. MADE IN THE USA Case qty-18 TOLERANCE +-(ML):0.25OAL (MM):230COLOR CODE: White Graduation: 0.5