Learn More
Bellco Glass, Inc. Shortie Pipet Large Orfice,25x.5mL Cotton Plug Top
SDP
Catalog No. 501944640
Encompass_Preferred
Bellco manufactures all precision pipettes from type 1 class a Borosilicate glass. All bellco pipettes feature tempered tips and permanent silk screened graduations to withstand repeated usage and alkali washings. 3mm orifice for handling tissue homogenizer. MADE IN THE USA Case qty-18 TOLERANCE +-(ML):0.25OAL (MM):225COLOR CODE: White Graduation: 0.5
Catalog No. 50-194-4640 Supplier Bellco Glass, Inc. Supplier No. 122825050
Bellco manufactures all precision pipettes from type 1 class a Borosilicate glass. All bellco pipettes feature tempered tips and permanent silk screened graduations to withstand repeated usage and alkali washings. 3mm orifice for handling tissue homogenizer. MADE IN THE USA Case qty-18 TOLERANCE +-(ML):0.25OAL (MM):225COLOR CODE: White Graduation: 0.5