Learn More
Research Products International Corp Shorty Serological Pipet, Sterile, Individually Wrapped, 10ml, Sterile, 150 per Case
SDP
Catalog No. 50998506
Encompass
Mini-Serological pipettes are only nine inches long. This shorter length provides a greater convenience when pipetting in tissue culture hoods and tight spaces. Pipettes made from transparent medical grade polystyrene and feature easy to read graduation, lint free plugs and color coded rings for quick identification. Pipettes are individually wrapped and sterile.
Catalog No. 50-998-506 Supplier Research Products International Corp Supplier No. 218401
Mini-Serological pipettes are only nine inches long. This shorter length provides a greater convenience when pipetting in tissue culture hoods and tight spaces. Pipettes made from transparent medical grade polystyrene and feature easy to read graduation, lint free plugs and color coded rings for quick identification. Pipettes are individually wrapped and sterile.