Learn More
Bellco Glass, Inc. Shorty Plain Top Pip,5x.1mL-Bulk
SDP
Catalog No. 501944582
Encompass_Preferred
Bellco manufactures all precision pipettes from type 1 class a Borosilicate glass. All bellco pipettes feature tempered tips and permanent silk screened graduations to withstand repeated usage and alkali washings. Shorter length facilitates use use under biological hoods. MADE IN THE USA Case Qty: 150 TOLERANCE +-(ML):0.8OAL (MM):160COLOR CODE: Blue Graduation: 0.1
Catalog No. 50-194-4582 Supplier Bellco Glass, Inc. Supplier No. 120405010
Bellco manufactures all precision pipettes from type 1 class a Borosilicate glass. All bellco pipettes feature tempered tips and permanent silk screened graduations to withstand repeated usage and alkali washings. Shorter length facilitates use use under biological hoods. MADE IN THE USA Case Qty: 150 TOLERANCE +-(ML):0.8OAL (MM):160COLOR CODE: Blue Graduation: 0.1