Description
- High-grade polystyrene
- Clear, easily legible graduation
- Colored vertical stripe for ease in reading the volume
- With synthetic fibrous filter material for protection against
Specifications
Specifications
| Description | Shape: Shorty |
| Material | Polystyrene |
| Sterility | Sterile |
| Graduated | Yes |
| Graduations | 1/10 |
| Packaging Type | Individually packed in plastic |
| Volume (Metric) | 5 mL |
| Type | Serological Pipet |