Learn More
Greiner Bio-One Shorty Sterile Serological Pipets
Catalog No. 07000361 Shop All Greiner Bio-One Products
Change view
Click to view available options
Volume (Metric):
10 mL
Graduations:
2/10
Catalog No. Volume (Metric) Graduations
07-000-361 10 mL 2/10
1 options
Catalog No. 07-000-361 Supplier Greiner Bio-One Supplier No. 607190
Drip-free pipetting design with high optical clarity and maximum accuracy.
  • High-grade polystyrene
  • Clear, easily legible graduation
  • Colored vertical stripe for ease in reading the volume
  • With synthetic fibrous filter material for protection against
TRUSTED_SUSTAINABILITY
Specifications
Description Shape: Shorty
Material Polystyrene
Sterility Sterile
Graduated Yes
Graduations 2/10
Packaging Type Individually packed in plastic
Volume (Metric) 10 mL
Type Serological Pipet