Learn More
Sigma Aldrich Fine Chemicals Biosciences Shorty VialsR volume 2 mL
Catalog No. 502901165
Encompass
Vial only. Autoclavable borosilicate USP Type 1 glass. Select caps and septa according to application.
Catalog No. 50-290-1165 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z106372200EA
Vial only. Autoclavable borosilicate USP Type 1 glass. Select caps and septa according to application.