Learn More
Northern Safety SHOULDER LENGTH PE GLOVE 100PK
SDP
Catalog No. NC0607065
Encompass
Economical, Multi-Purpose Gloves Sheer poly film offers excellent sense of touch without the worry of tearing or ripping Powder-free/sulphur-free surface won't mar delicate materials Ambidextrous design fits either hand for easy distribution and wear Light duty protection is ideal for food processing, small parts handling, janitorial, and maintenance applications Made with FDA accepted materials 100 per box 10 boxes per case
This item is not returnable. View return policy
Catalog No. NC0607065 Supplier Northern Safety Supplier No. 16747
Economical, Multi-Purpose Gloves Sheer poly film offers excellent sense of touch without the worry of tearing or ripping Powder-free/sulphur-free surface won't mar delicate materials Ambidextrous design fits either hand for easy distribution and wear Light duty protection is ideal for food processing, small parts handling, janitorial, and maintenance applications Made with FDA accepted materials 100 per box 10 boxes per case