Learn More
MSC SHOWA Atlas KV300 Gloves, Kevlar, Latex, Cut A3, 1 Pair
Catalog No. 5000675522
Encompass
The SHOWA Atlas KV300 gloves are cut, puncture, and abrasion-resistant, featuring a 10-gauge seamless Kevlar liner and a latex coating on the palm and fingertips for a rough grip.
  • Seamless knit construction
  • Liner made with Kevlar
  • Palm-dipped natural rubber coating
  • Rough grip surface
  • ANSI/ISEA cut resistance level A3
  • ANSI/ISEA puncture resistance level 5
  • ANSI/ISEA abrasion resistance level 4
  • Features a 10-gauge knit
  • Size 2X-Large (11)
  • Overall length 10-1/2 in
  • Low thermal conductivity
Catalog No. 50-006-75522 Supplier MSC Supplier No. 91309237
The SHOWA Atlas KV300 gloves are cut, puncture, and abrasion-resistant, featuring a 10-gauge seamless Kevlar liner and a latex coating on the palm and fingertips for a rough grip.
  • Seamless knit construction
  • Liner made with Kevlar
  • Palm-dipped natural rubber coating
  • Rough grip surface
  • ANSI/ISEA cut resistance level A3
  • ANSI/ISEA puncture resistance level 5
  • ANSI/ISEA abrasion resistance level 4
  • Features a 10-gauge knit
  • Size 2X-Large (11)
  • Overall length 10-1/2 in
  • Low thermal conductivity