Learn More
MSC Showa Chemical Gloves, Viton, 12 mil, 12 in, 1 Pair
Catalog No. 5000793688
Encompass
SHOWA 892 gloves are forged with fluoroelastomer rubber, offering superior resistance to highly corrosive acids and designed for handling aliphatic and aromatic hydrocarbons such as Benzene, Toluene, and Xylene.
  • Superior resistance to highly corrosive acids
  • Designed for handling aliphatic and aromatic hydrocarbons
  • Made from Viton® (fluoroelastomer rubber)
  • Features 12 mil thickness
  • Offers 12 in length
  • Black color
  • Smooth finish
  • Industrial grade
Catalog No. 50-007-93688 Supplier MSC Supplier No. 66004458
SHOWA 892 gloves are forged with fluoroelastomer rubber, offering superior resistance to highly corrosive acids and designed for handling aliphatic and aromatic hydrocarbons such as Benzene, Toluene, and Xylene.
  • Superior resistance to highly corrosive acids
  • Designed for handling aliphatic and aromatic hydrocarbons
  • Made from Viton® (fluoroelastomer rubber)
  • Features 12 mil thickness
  • Offers 12 in length
  • Black color
  • Smooth finish
  • Industrial grade