Learn More
MSC SHOWA Chemical Resistant Gloves, Butyl, 0.35 mm, 2X-Large, 1 Pair
Catalog No. 5000687727
Encompass
The SHOWA 874 chemical resistant glove offers superior protection against highly corrosive acids, ketones, and esters. Butyl rubber provides the highest permeation resistance to gases and water vapors of any material used to make gloves. A smooth surface grip provides unmatched tactility and performance.
  • Unlined
  • Rolled cuff
  • Smooth grip
  • Unsupported
  • Butyl rubber construction
  • Acid-resistant
  • Impermeable to liquids and gases
  • Water-resistant
Catalog No. 50-006-87727 Supplier MSC Supplier No. 66004391
The SHOWA 874 chemical resistant glove offers superior protection against highly corrosive acids, ketones, and esters. Butyl rubber provides the highest permeation resistance to gases and water vapors of any material used to make gloves. A smooth surface grip provides unmatched tactility and performance.
  • Unlined
  • Rolled cuff
  • Smooth grip
  • Unsupported
  • Butyl rubber construction
  • Acid-resistant
  • Impermeable to liquids and gases
  • Water-resistant