Learn More
MSC SHOWA Chemical Resistant Gloves, Butyl, 14 mil, Large, 1 Pair
Catalog No. 5000687726
Encompass
This chemical resistant glove offers superior protection against highly corrosive acids, ketones, and esters. Butyl rubber provides the highest permeation resistance to gases and water vapors of any material used to make gloves, while a smooth surface grip ensures unmatched tactility and performance.
  • Unsupported design for dexterity in light-to-medium chemical applications
  • Unlined for direct feel
  • Rolled cuff for easy donning and doffing
  • Smooth grip surface
  • Made from impermeable butyl rubber
  • Acid-resistant and water-resistant
Catalog No. 50-006-87726 Supplier MSC Supplier No. 66004375
This chemical resistant glove offers superior protection against highly corrosive acids, ketones, and esters. Butyl rubber provides the highest permeation resistance to gases and water vapors of any material used to make gloves, while a smooth surface grip ensures unmatched tactility and performance.
  • Unsupported design for dexterity in light-to-medium chemical applications
  • Unlined for direct feel
  • Rolled cuff for easy donning and doffing
  • Smooth grip surface
  • Made from impermeable butyl rubber
  • Acid-resistant and water-resistant