Learn More
MSC SHOWA Chemical Resistant Gloves, Large, 16 mil, Polyvinylchloride, 1 Pair
Catalog No. 5000765798
Encompass
These gloves feature a polyvinylchloride-coated cotton construction and a rough grip surface, designed to provide chemical resistance and enhanced handling.
  • Polyvinylchloride-coated cotton construction
  • Rough grip surface
  • Provides chemical resistance
  • Enhanced handling
Catalog No. 50-007-65798 Supplier MSC Supplier No. 70756812
These gloves feature a polyvinylchloride-coated cotton construction and a rough grip surface, designed to provide chemical resistance and enhanced handling.
  • Polyvinylchloride-coated cotton construction
  • Rough grip surface
  • Provides chemical resistance
  • Enhanced handling