Learn More
MSC SHOWA Chemical Resistant Gloves, Large, Viton, 12.00 mil, 1 Pair
Catalog No. 5000766700
Encompass
These chemical resistant gloves are constructed from Viton, offering protection against a range of chemicals. Designed for tasks requiring dexterity, they feature an unsupported, unlined design with a smooth grip and gauntlet cuff.
  • Made from 100% fluoroelastomer
  • Thickness of 12.00 mil
  • Resists toluene, aromatic hydrocarbons, benzene, xylene, highly corrosive acids, and aliphatics
  • Certified to EN 388:2016 1010X, EN 420:2003+A1:2009, EN ISO 374-5:2016 VIRUS, CAT III CE 0299, and EN ISO 374-1:2016/Type B DFL
  • Size large, numeric size 9
  • Overall length of 12.00 in.
Catalog No. 50-007-66700 Supplier MSC Supplier No. 66004474
These chemical resistant gloves are constructed from Viton, offering protection against a range of chemicals. Designed for tasks requiring dexterity, they feature an unsupported, unlined design with a smooth grip and gauntlet cuff.
  • Made from 100% fluoroelastomer
  • Thickness of 12.00 mil
  • Resists toluene, aromatic hydrocarbons, benzene, xylene, highly corrosive acids, and aliphatics
  • Certified to EN 388:2016 1010X, EN 420:2003+A1:2009, EN ISO 374-5:2016 VIRUS, CAT III CE 0299, and EN ISO 374-1:2016/Type B DFL
  • Size large, numeric size 9
  • Overall length of 12.00 in.