Learn More
MSC SHOWA Chemical Resistant Gloves, Medium, 14 mil, Butyl, 1 Pair
Catalog No. 5000792453
Encompass
These chemical resistant gloves are designed for tasks requiring dexterity and protection against a range of chemicals. Made from butyl, they are suitable for light-to-medium chemical applications.
  • Unsupported design for dexterity and economical use
  • Protection against ketones, esters, and highly corrosive acids
  • Unlined with a rolled cuff style
  • Black color and rough grip surface
  • Meets EN 388:2016 0010X, EN 420:2003+A1:2009, EN ISO 374-5:2016 VIRUS, CAT III CE 0299, and EN ISO 374-1:2016/Type B BIK standards
Catalog No. 50-007-92453 Supplier MSC Supplier No. 66004417
These chemical resistant gloves are designed for tasks requiring dexterity and protection against a range of chemicals. Made from butyl, they are suitable for light-to-medium chemical applications.
  • Unsupported design for dexterity and economical use
  • Protection against ketones, esters, and highly corrosive acids
  • Unlined with a rolled cuff style
  • Black color and rough grip surface
  • Meets EN 388:2016 0010X, EN 420:2003+A1:2009, EN ISO 374-5:2016 VIRUS, CAT III CE 0299, and EN ISO 374-1:2016/Type B BIK standards