Learn More
MSC SHOWA Chemical Resistant Gloves, Neoprene-Coated, Supported, Size Large, 1 Pair
Catalog No. 5000677337
Encompass
These chemical resistant gloves are constructed from multi-dipped, premium-grade neoprene, offering high durability, oil protection, and wet grip. Designed to protect against physical hazards like cuts and abrasion, they feature a soft cotton liner for enhanced comfort and tactile precision.
  • Cut and chemical resistant
  • Neoprene material with neoprene coating
  • Supported design with flannel lining
  • Resistant to solvents, acids, oils, and general chemicals
  • Smooth grip surface and knit cuff
  • ANSI/ISEA Abrasion Resistance Level 3
  • ANSI/ISEA Cut Resistance Level A3
  • FDA Approved
  • EN 388:2016+A1:2018 (2111X) certified
Catalog No. 50-006-77337 Supplier MSC Supplier No. 00255315
These chemical resistant gloves are constructed from multi-dipped, premium-grade neoprene, offering high durability, oil protection, and wet grip. Designed to protect against physical hazards like cuts and abrasion, they feature a soft cotton liner for enhanced comfort and tactile precision.
  • Cut and chemical resistant
  • Neoprene material with neoprene coating
  • Supported design with flannel lining
  • Resistant to solvents, acids, oils, and general chemicals
  • Smooth grip surface and knit cuff
  • ANSI/ISEA Abrasion Resistance Level 3
  • ANSI/ISEA Cut Resistance Level A3
  • FDA Approved
  • EN 388:2016+A1:2018 (2111X) certified