Learn More
MSC SHOWA Chemical Resistant Gloves, Neoprene/Latex, 26 mil, Medium, 1 Pair
Catalog No. 5000795847
Encompass
These chemical protection gloves are flexible, comfortable, and resistant to a broad range of chemicals. Engineered with a neoprene-over-natural rubber construction and tractor tread grip, they provide maximum precision, sensitivity, and defense. A cotton flocked liner helps maintain a comfortable temperature within the glove.
  • Flexible and comfortable for extended wear
  • Increased sensitivity for precision tasks
  • Resistant to a broad range of chemicals
  • Neoprene-over-natural rubber construction
  • Embossed grip for enhanced handling
  • Supported design for additional strength
  • Cotton flocked liner for temperature regulation
Catalog No. 50-007-95847 Supplier MSC Supplier No. 00255620
These chemical protection gloves are flexible, comfortable, and resistant to a broad range of chemicals. Engineered with a neoprene-over-natural rubber construction and tractor tread grip, they provide maximum precision, sensitivity, and defense. A cotton flocked liner helps maintain a comfortable temperature within the glove.
  • Flexible and comfortable for extended wear
  • Increased sensitivity for precision tasks
  • Resistant to a broad range of chemicals
  • Neoprene-over-natural rubber construction
  • Embossed grip for enhanced handling
  • Supported design for additional strength
  • Cotton flocked liner for temperature regulation