Learn More
MSC SHOWA Chemical Resistant Gloves, Polyvinylchloride, Electrostatic Dissipative, Large, 1 Pair
Catalog No. 5000769492
Encompass
These SHOWA Atlas chemical resistant gloves are designed for applications requiring protection against chemicals and electrostatic discharge. They feature a durable polyvinylchloride coating and a rough grip for enhanced handling.
  • Polyvinylchloride material
  • Large glove size
  • Polyvinylchloride coating
  • Rough grip surface
  • Gauntlet cuff style
  • Cotton lining
  • Electrostatic dissipative
  • Overall length of 12.00 in
Catalog No. 50-007-69492 Supplier MSC Supplier No. 40752529
These SHOWA Atlas chemical resistant gloves are designed for applications requiring protection against chemicals and electrostatic discharge. They feature a durable polyvinylchloride coating and a rough grip for enhanced handling.
  • Polyvinylchloride material
  • Large glove size
  • Polyvinylchloride coating
  • Rough grip surface
  • Gauntlet cuff style
  • Cotton lining
  • Electrostatic dissipative
  • Overall length of 12.00 in