Learn More
MSC SHOWA Cut and Puncture-Resistant Sleeve, HPPE, ANSI A3, 1 ea
Catalog No. 5000673051
Encompass
This seamless sleeve is designed for protection against cuts and punctures. Made from HPPE material, it features a thumb hole and meets ANSI Cut A3 standards.
  • Made from HPPE material
  • Seamless construction
  • Features a thumb hole
  • Meets ANSI Cut A3 standards
  • CE and ANSI/ISEA compliant
  • Suitable for HVAC, food packing, handling glass, bottling, windows, and beverage handling
  • Super stretch for comfort and flexibility
Catalog No. 50-006-73051 Supplier MSC Supplier No. 30191167
This seamless sleeve is designed for protection against cuts and punctures. Made from HPPE material, it features a thumb hole and meets ANSI Cut A3 standards.
  • Made from HPPE material
  • Seamless construction
  • Features a thumb hole
  • Meets ANSI Cut A3 standards
  • CE and ANSI/ISEA compliant
  • Suitable for HVAC, food packing, handling glass, bottling, windows, and beverage handling
  • Super stretch for comfort and flexibility