Learn More
MSC SHOWA Cut-Resistant Gloves, Cotton/Latex, ANSI Cut A1, Small, 72 Pairs
Catalog No. 5000788701
Encompass
The SHOWA L66NFW Cut-Resistant Gloves are designed for utility applications, featuring a cotton primary material and a latex coating for enhanced grip and protection. These gloves provide a 3/4 dip and palm coating with a rough grip, ensuring durability and safety in various work environments.
  • Cotton flannel liner
  • Rough grip
  • Reinforced safety cuff
  • Palm-dipped natural rubber coating
Catalog No. 50-007-88701 Supplier MSC Supplier No. 62388681
The SHOWA L66NFW Cut-Resistant Gloves are designed for utility applications, featuring a cotton primary material and a latex coating for enhanced grip and protection. These gloves provide a 3/4 dip and palm coating with a rough grip, ensuring durability and safety in various work environments.
  • Cotton flannel liner
  • Rough grip
  • Reinforced safety cuff
  • Palm-dipped natural rubber coating