Learn More
MSC SHOWA Disposable Gloves, Nitrile, 4 mil, Biodegradable, Small, 100/Pk.
Catalog No. 5000793301
Encompass
The SHOWA 7500PF disposable gloves feature Eco Best Technology (EBT) for biodegradability, offering an environmentally conscious choice for laboratory and industrial use. These 4 mil, powder-free, and latex-free nitrile gloves provide increased dexterity and a textured grip, ensuring safety and comfort across a range of applications.
  • Biodegradable with Eco Best Technology
  • Flexible and ergonomic design
  • Latex-free and powder-free
  • Increased dexterity
  • Textured finish for enhanced grip
  • Rolled cuff prevents dirt entry
  • Made from 100% nitrile, 4 mil thickness
  • FDA approved and non-sterile
  • Certified to EN ISO 374-1/Type B (JKPT), EN ISO 374-5 (VIRUS), Cat III
  • Food contact certified
Catalog No. 50-007-93301 Supplier MSC Supplier No. 74882093
The SHOWA 7500PF disposable gloves feature Eco Best Technology (EBT) for biodegradability, offering an environmentally conscious choice for laboratory and industrial use. These 4 mil, powder-free, and latex-free nitrile gloves provide increased dexterity and a textured grip, ensuring safety and comfort across a range of applications.
  • Biodegradable with Eco Best Technology
  • Flexible and ergonomic design
  • Latex-free and powder-free
  • Increased dexterity
  • Textured finish for enhanced grip
  • Rolled cuff prevents dirt entry
  • Made from 100% nitrile, 4 mil thickness
  • FDA approved and non-sterile
  • Certified to EN ISO 374-1/Type B (JKPT), EN ISO 374-5 (VIRUS), Cat III
  • Food contact certified