Learn More
MSC SHOWA Disposable Gloves, Nitrile, 6 mil, Black, Powder-Free, 50/Box
Catalog No. 5000792811
Encompass
These single-use gloves are designed for comfort, highly tactile precision, and enhanced protection across various applications. Featuring a rolled cuff to prevent dirt entry and a thicker nitrile construction, they offer a greater barrier to hazards. This product incorporates advanced technology for accelerated biodegradation in biologically active landfills.
  • Ergonomic, form-fitting design
  • Lightweight and disposable
  • Powder-free, latex-free, and accelerator-free
  • Increased tactile sensitivity
  • Textured fingertips for improved grip
  • Rolled cuff for dirt prevention
  • Biodegradable with Eco Best Technology® (EBT)
  • Constructed from 100% nitrile
  • Low modulus formulation for comfort
  • Reduced risk of type IV allergies
Catalog No. 50-007-92811 Supplier MSC Supplier No. 40627945
These single-use gloves are designed for comfort, highly tactile precision, and enhanced protection across various applications. Featuring a rolled cuff to prevent dirt entry and a thicker nitrile construction, they offer a greater barrier to hazards. This product incorporates advanced technology for accelerated biodegradation in biologically active landfills.
  • Ergonomic, form-fitting design
  • Lightweight and disposable
  • Powder-free, latex-free, and accelerator-free
  • Increased tactile sensitivity
  • Textured fingertips for improved grip
  • Rolled cuff for dirt prevention
  • Biodegradable with Eco Best Technology® (EBT)
  • Constructed from 100% nitrile
  • Low modulus formulation for comfort
  • Reduced risk of type IV allergies