Learn More
MSC SHOWA Disposable Gloves, Nitrile, 6 mil, Medium, Powder-Free, 50/Pk.
Catalog No. 5000795318
Encompass
These disposable gloves are made of durable nitrile and are suitable for various applications, including food handling. They are FDA approved and designed to provide protection and comfort during use.
  • Thickness of 6 mil
  • Powder-free
  • Ambidextrous design
  • Extended, rolled cuff
  • Smooth grip surface
  • FDA approved for food contact
  • Non-sterile
  • Blue color
  • Length of 11.0 in
  • Package quantity of 50
Catalog No. 50-007-95318 Supplier MSC Supplier No. 33756529
These disposable gloves are made of durable nitrile and are suitable for various applications, including food handling. They are FDA approved and designed to provide protection and comfort during use.
  • Thickness of 6 mil
  • Powder-free
  • Ambidextrous design
  • Extended, rolled cuff
  • Smooth grip surface
  • FDA approved for food contact
  • Non-sterile
  • Blue color
  • Length of 11.0 in
  • Package quantity of 50