Learn More
MSC SHOWA Disposable Gloves, Nitrile, 6 mil, Powder-Free, Large, 50 Pack
Catalog No. 5000795627
Encompass
These single-use nitrile gloves offer comfortable and highly tactile precision with high-capacity protection for a wide range of applications. A rolled cuff prevents dirt from entering, and the thicker nitrile construction provides an enhanced barrier against hazards. Featuring Eco Best Technology® (EBT), these gloves are engineered for accelerated biodegradation in biologically active landfills.
  • Ergonomic, form-fitting, and lightweight design
  • Powder-free and latex-free
  • Disposable for convenience and hygiene
  • Enhanced sensitivity with textured fingertips for improved grip
  • Biodegradable with Eco Best Technology® (EBT)
  • Rolled cuff prevents ingress of dirt
  • Static dissipative and non-sterile
  • Approved for food contact (FDA 21 CFR Part 177)
  • Accelerator-free construction for reduced skin irritation
Catalog No. 50-007-95627 Supplier MSC Supplier No. 40627937
These single-use nitrile gloves offer comfortable and highly tactile precision with high-capacity protection for a wide range of applications. A rolled cuff prevents dirt from entering, and the thicker nitrile construction provides an enhanced barrier against hazards. Featuring Eco Best Technology® (EBT), these gloves are engineered for accelerated biodegradation in biologically active landfills.
  • Ergonomic, form-fitting, and lightweight design
  • Powder-free and latex-free
  • Disposable for convenience and hygiene
  • Enhanced sensitivity with textured fingertips for improved grip
  • Biodegradable with Eco Best Technology® (EBT)
  • Rolled cuff prevents ingress of dirt
  • Static dissipative and non-sterile
  • Approved for food contact (FDA 21 CFR Part 177)
  • Accelerator-free construction for reduced skin irritation