Learn More
MSC SHOWA Nitrile Gloves, 2.5 mil, Powder-Free, Textured Fingertips, Small, 100/Pack
Catalog No. 5000795084
Encompass
The 7503PF disposable glove is powder and latex-free, designed for single-use applications. It features a SafeDoff textured tab for safer and faster doffing after handling messy or oily substances, or when exposed to hazardous materials. The textured surface grip enhances safety and ease during wet work.
  • Constructed from 100% nitrile
  • Ergonomic design
  • SafeDoff textured tab for easier doffing
  • Textured fingertips for enhanced grip
  • Touch screen compatible
  • Powder and latex-free
Catalog No. 50-007-95084 Supplier MSC Supplier No. 39258140
The 7503PF disposable glove is powder and latex-free, designed for single-use applications. It features a SafeDoff textured tab for safer and faster doffing after handling messy or oily substances, or when exposed to hazardous materials. The textured surface grip enhances safety and ease during wet work.
  • Constructed from 100% nitrile
  • Ergonomic design
  • SafeDoff textured tab for easier doffing
  • Textured fingertips for enhanced grip
  • Touch screen compatible
  • Powder and latex-free