Learn More
MSC SHOWA S-TEX 300 Gloves, Cut A4, Large, 1 Pair
Catalog No. 5000768019
Encompass
These SHOWA S-TEX 300 series gloves are designed for superior cut, puncture, and abrasion resistance in demanding work environments. They combine advanced materials with an ergonomic design for enhanced protection and comfort.
  • Cut A4, Puncture 4, and Abrasion 4 ratings for high-level protection
  • Constructed from polyester and stainless steel for durability
  • Latex coating on palm and fingertips provides a rough, secure grip
  • Seamless knit lining ensures comfort and prevents irritation
  • Ergonomic design for extended wear
  • Suitable for applications requiring high dexterity and protection
Catalog No. 50-007-68019 Supplier MSC Supplier No. 51640951
These SHOWA S-TEX 300 series gloves are designed for superior cut, puncture, and abrasion resistance in demanding work environments. They combine advanced materials with an ergonomic design for enhanced protection and comfort.
  • Cut A4, Puncture 4, and Abrasion 4 ratings for high-level protection
  • Constructed from polyester and stainless steel for durability
  • Latex coating on palm and fingertips provides a rough, secure grip
  • Seamless knit lining ensures comfort and prevents irritation
  • Ergonomic design for extended wear
  • Suitable for applications requiring high dexterity and protection