Learn More
MSC SHOWA Work Gloves, 406 Series, Medium, Latex-Coated, 1 Pair
Catalog No. 5000673100
Encompass
These general purpose work gloves feature a seamless nylon and polyester liner with a full latex coating. Designed for various tasks, these gloves provide rough grip and protection against cold temperatures.
  • Constructed from durable nylon and polyester for comfort
  • Full latex coating ensures enhanced grip and protection
  • Rough grip surface facilitates secure handling in diverse conditions
  • Seamless design reduces irritation for prolonged wear
  • Gauntlet cuff style offers extended wrist coverage
  • Ideal for plumbing, brick laying, pool and pond maintenance, washing and cleaning, and shipping and receiving
  • Offers cold protection down to -20°F (-28.9°C)
  • Unisex sizing ensures versatile use
Catalog No. 50-006-73100 Supplier MSC Supplier No. 31250244
These general purpose work gloves feature a seamless nylon and polyester liner with a full latex coating. Designed for various tasks, these gloves provide rough grip and protection against cold temperatures.
  • Constructed from durable nylon and polyester for comfort
  • Full latex coating ensures enhanced grip and protection
  • Rough grip surface facilitates secure handling in diverse conditions
  • Seamless design reduces irritation for prolonged wear
  • Gauntlet cuff style offers extended wrist coverage
  • Ideal for plumbing, brick laying, pool and pond maintenance, washing and cleaning, and shipping and receiving
  • Offers cold protection down to -20°F (-28.9°C)
  • Unisex sizing ensures versatile use