Learn More
MSC SHOWA Work Gloves, Nitrile-Coated, Nylon and Polyester, Medium, 1 Pair
Catalog No. 5000802312
Encompass
These general purpose work gloves are designed with a nitrile coating on the palm and fingertips for enhanced grip and durability. Constructed from a blend of nylon and polyester, these 9 in long gloves are suitable for various applications, offering both protection and comfort.
  • Nitrile coating on palm and fingertips
  • Constructed from polyester and nylon materials
  • Foam grip surface for improved handling
  • Elastic cuff style for a secure fit
  • Seamless design for comfort
  • Certified to CAT II and EN 388:2016 4121X standards
  • Overall length of 9 in
  • Full fingered design
  • Suitable for construction, automotive, and petrochemical applications
Catalog No. 50-008-02312 Supplier MSC Supplier No. 51627016
These general purpose work gloves are designed with a nitrile coating on the palm and fingertips for enhanced grip and durability. Constructed from a blend of nylon and polyester, these 9 in long gloves are suitable for various applications, offering both protection and comfort.
  • Nitrile coating on palm and fingertips
  • Constructed from polyester and nylon materials
  • Foam grip surface for improved handling
  • Elastic cuff style for a secure fit
  • Seamless design for comfort
  • Certified to CAT II and EN 388:2016 4121X standards
  • Overall length of 9 in
  • Full fingered design
  • Suitable for construction, automotive, and petrochemical applications