Learn More
3M™ RBE-SC Shower Cover
Catalog No. 19070841 Shop All 3M Products
Change view
Click to view available options
Quantity:
2/Pk.
Catalog No. Quantity
19-070-841 2/Pk.
1 options
Catalog No. 19-070-841 Supplier 3M™ Supplier No. 7000126950
  • $190.09
    $133.65 / Case of 1
    Save $56.44
    30% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Shower cover helps to protect the RBE PAPR assemblies RBE-40 and RBE-57 from moisture.
  • Protects RBE PAPR Assemblies RBE-40 and RBE-57 from moisture, but does not eliminate moisture entry completely
  • For use by first responders, law enforcement, military, and healthcare professionals
  • 3M Catalog# RBE-SC
TRUSTED_SUSTAINABILITY
Specifications
Disposable Reusable
For Use With (Equipment) RBE-40 and RBE-57 PAPR Assemblies
Model RBE-SC
Quantity 2/Pk.