Learn More
Grainger Haws Shower and Eyewash, Barrier-Free, Recessed, Stainless Steel
Catalog No. 502792145
Encompass_Preferred
The AXION MSR™ barrier-free combination shower and eye/face wash is designed for environments where immediate flushing of the eyes, face, and body is required following exposure to injurious or corrosive materials. This recessed unit features an 18 gauge, Type 304 stainless steel cabinet with a smudge-resistant bead-blast finish and a pull-down design that accommodates all users, including those in wheelchairs. It utilizes inverted directional laminar flow technology to provide a safe, effective decontamination experience.
  • Features AXION MSR hydrodynamic stainless steel showerhead for uniform water distribution
  • Equipped with AXION MSR eye/face wash head to sweep contaminants away from the nasal cavity
  • Includes polished chrome-plated brass pull-down valve with easy-access in-line strainer
  • Certified by CSA to meet ANSI/ISEA Z358.1 standard
  • Includes universal sign and inspection tag
  • <
Catalog No. 50-279-2145 Supplier Grainger Supplier No. 16D407
The AXION MSR™ barrier-free combination shower and eye/face wash is designed for environments where immediate flushing of the eyes, face, and body is required following exposure to injurious or corrosive materials. This recessed unit features an 18 gauge, Type 304 stainless steel cabinet with a smudge-resistant bead-blast finish and a pull-down design that accommodates all users, including those in wheelchairs. It utilizes inverted directional laminar flow technology to provide a safe, effective decontamination experience.
  • Features AXION MSR hydrodynamic stainless steel showerhead for uniform water distribution
  • Equipped with AXION MSR eye/face wash head to sweep contaminants away from the nasal cavity
  • Includes polished chrome-plated brass pull-down valve with easy-access in-line strainer
  • Certified by CSA to meet ANSI/ISEA Z358.1 standard
  • Includes universal sign and inspection tag
  • <