Learn More
Grainger Haws Barrier-Free Combination Shower and Eye/Face Wash, Stainless Steel, Recessed Wall-Mount, 1 ea
Catalog No. 502791448
Encompass_Preferred
This barrier-free combination shower and eye/face wash system is designed for safety in work areas where exposure to injurious or corrosive materials is possible. The unit features a fully recessed, wall-mounted cabinet constructed from 18 gauge Type 304 stainless steel with a smooth electro-polished finish for long-lasting durability. The system includes an AXION® MSR eye/face wash head with inverted directional laminar flow to sweep contaminants away from the nasal cavity, along with a hydrodynamic stainless steel showerhead that ensures equal water distribution. This unit is CSA certified to meet the ANSI/ISEA Z358.1 standard.
  • Wheelchair accessible pull-down design for universal use
  • Inverted directional laminar flow eye/face wash head
  • Hydrodynamic stainless steel showerhead for uniform coverage
  • Durable 18 gauge Type 304 stainless steel recessed cabinet
  • Includes brass pipe and fittings
Catalog No. 50-279-1448 Supplier Grainger Supplier No. 16D408
This barrier-free combination shower and eye/face wash system is designed for safety in work areas where exposure to injurious or corrosive materials is possible. The unit features a fully recessed, wall-mounted cabinet constructed from 18 gauge Type 304 stainless steel with a smooth electro-polished finish for long-lasting durability. The system includes an AXION® MSR eye/face wash head with inverted directional laminar flow to sweep contaminants away from the nasal cavity, along with a hydrodynamic stainless steel showerhead that ensures equal water distribution. This unit is CSA certified to meet the ANSI/ISEA Z358.1 standard.
  • Wheelchair accessible pull-down design for universal use
  • Inverted directional laminar flow eye/face wash head
  • Hydrodynamic stainless steel showerhead for uniform coverage
  • Durable 18 gauge Type 304 stainless steel recessed cabinet
  • Includes brass pipe and fittings