This free-standing combination safety shower and eye/face wash station features an AXION® MSR design to provide medically superior emergency response. It is engineered to deliver uniform drenching and consistent flow, sweeping contaminants away from vulnerable areas, including the nasal cavity. The unit is constructed for durability in industrial environments, featuring a green corrosion-resistant epoxy-coated galvanized steel pipe and an 11 in. (27.9 cm) ABS plastic bowl. It is pre-built and pressure-tested to ensure reliable performance and facilitate easier installation.
- Features AXION® MSR eye/face wash head with inverted directional laminar flow and a 3.7 gpm flow control
- Includes a hydrodynamic ABS plastic drench showerhead with 20 gpm flow control for uniform drenching
- Equipped with chrome-plated brass stay-open ball valves featuring stainless steel balls and stems
- Includes a mesh in-