This floor-mounted, combination shower and eye/face wash system is designed for use in environments where individuals may be exposed to injurious or corrosive materials. The unit features AXION® MSR technology, which utilizes an inverted directional laminar flow in the eye/face wash head to effectively sweep contaminants away from the nasal cavity. The showerhead uses a hydrodynamic design to provide equal water distribution throughout the flow footprint. The unit is constructed with durable, corrosion-resistant materials and includes integrated safety features for emergency response.
- Features AXION® MSR hydrodynamic eye/face wash and drench showerheads.
- Constructed with 1-1/4 in. IPS Schedule 40 hot-dipped galvanized steel pipe and fittings.
- Includes a 27.9 cm (11 in.) stainless steel bowl and a floor flange coated with green epoxy.
- Equipped with chrome-plated brass ball valves featuring st