Learn More
Chemglass Life Sciences Chemglass Shower Head Adapter for 20 L Gas Scrubber, 100 mm Column
SDP
Catalog No. 502733576
Encompass_Preferred
Shower head adapter designed as a replacement component for 20 L gas scrubbers with a 100 mm column. It mounts to the top of the exhaust gas outlet column and disperses scrubbing fluid through a shower head to improve liquid-gas contact and absorption efficiency. Sold individually.
  • Attaches to the top of the exhaust gas outlet column.
  • Disperses scrubbing fluid through a shower head for improved contact and absorption.
  • Designed for use with 100 mm columns in 20 L gas scrubbers.
  • Sold individually for replacement or maintenance.
Catalog No. 50-273-3576 Supplier Chemglass Life Sciences Supplier No. CG183055
Shower head adapter designed as a replacement component for 20 L gas scrubbers with a 100 mm column. It mounts to the top of the exhaust gas outlet column and disperses scrubbing fluid through a shower head to improve liquid-gas contact and absorption efficiency. Sold individually.
  • Attaches to the top of the exhaust gas outlet column.
  • Disperses scrubbing fluid through a shower head for improved contact and absorption.
  • Designed for use with 100 mm columns in 20 L gas scrubbers.
  • Sold individually for replacement or maintenance.