Learn More
Grainger Emergency Shower Tester, Vinyl, For Use With Emergency Shower Stations, 15 in Width
Catalog No. NC0105912
Encompass_Preferred
Keep this emergency shower tester handy to check water flow weekly in your workplace emergency showers. Use the shower tester to ensure emergency shower operates properly. Vinyl sleeve directs water into a plastic pail so you don’t have to get out a ladder, mop up or get wet. Plastic bucket included.
Catalog No. NC0105912 Supplier Grainger Supplier No. 8ND03
Keep this emergency shower tester handy to check water flow weekly in your workplace emergency showers. Use the shower tester to ensure emergency shower operates properly. Vinyl sleeve directs water into a plastic pail so you don’t have to get out a ladder, mop up or get wet. Plastic bucket included.