Learn More
Grainger Haws AXION MSR Combination Shower and Eye/Face Wash, Type 304 Stainless Steel, Fully Recessed, 1 ea
Catalog No. 502791991
Encompass_Preferred
This barrier-free combination shower and eye/face wash system is designed for emergency use where rapid, effective flushing of the eyes, face, and body is required. The unit features an 18-gauge, type 304 stainless steel, fully recessed wall-mounted cabinet for a streamlined, accessible design suitable for wheelchair users. It utilizes advanced, medically superior response technology to provide consistent, comfortable flow during emergencies.
  • Features the AXION® MSR eye/face wash head with inverted directional laminar flow that clears contaminants away from the nasal cavity
  • Includes a hydrodynamic design stainless steel showerhead that ensures equal water distribution across the flow footprint
  • Equipped with an 18-gauge, type 304 stainless steel deep-drawn electro-polished cabinet for durability
  • Includes a polished chrome-plated brass pull-down eye/face wash valve with an easy-access in-line stra
Catalog No. 50-279-1991 Supplier Grainger Supplier No. 16D409
This barrier-free combination shower and eye/face wash system is designed for emergency use where rapid, effective flushing of the eyes, face, and body is required. The unit features an 18-gauge, type 304 stainless steel, fully recessed wall-mounted cabinet for a streamlined, accessible design suitable for wheelchair users. It utilizes advanced, medically superior response technology to provide consistent, comfortable flow during emergencies.
  • Features the AXION® MSR eye/face wash head with inverted directional laminar flow that clears contaminants away from the nasal cavity
  • Includes a hydrodynamic design stainless steel showerhead that ensures equal water distribution across the flow footprint
  • Equipped with an 18-gauge, type 304 stainless steel deep-drawn electro-polished cabinet for durability
  • Includes a polished chrome-plated brass pull-down eye/face wash valve with an easy-access in-line stra