Learn More
Grainger Haws Barrier-Free Combination Shower and Eye/Face Wash, Stainless Steel
Catalog No. 502791508
Encompass_Preferred
This barrier-free, ceiling-mounted combination shower and recessed eye/face wash station is designed for use in laboratory environments where individuals may be exposed to hazardous materials. Featuring a durable 18-gauge, type 304 stainless steel cabinet with an electro-polished finish, this unit is built for long-term reliability. The integrated pull-down design and daylight drain feature ensure accessibility for all users, including those in wheelchairs, and allow for installation where in-wall drainage is unavailable.
  • Features an inverted directional laminar flow eye/face wash head to effectively sweep contaminants away from the nasal cavity.
  • Utilizes a hydrodynamic stainless steel showerhead for equal water distribution throughout the flow footprint.
  • Compliant with ANSI/ISEA Z358.1 standards for emergency eyewash and shower equipment.
  • Includes a polished chrome-plated brass pull-down eye/face w
Catalog No. 50-279-1508 Supplier Grainger Supplier No. 16D410
This barrier-free, ceiling-mounted combination shower and recessed eye/face wash station is designed for use in laboratory environments where individuals may be exposed to hazardous materials. Featuring a durable 18-gauge, type 304 stainless steel cabinet with an electro-polished finish, this unit is built for long-term reliability. The integrated pull-down design and daylight drain feature ensure accessibility for all users, including those in wheelchairs, and allow for installation where in-wall drainage is unavailable.
  • Features an inverted directional laminar flow eye/face wash head to effectively sweep contaminants away from the nasal cavity.
  • Utilizes a hydrodynamic stainless steel showerhead for equal water distribution throughout the flow footprint.
  • Compliant with ANSI/ISEA Z358.1 standards for emergency eyewash and shower equipment.
  • Includes a polished chrome-plated brass pull-down eye/face w