Learn More
Aviva Systems Biology Corp SHOX Antibody - N-terminal region (ARP33284_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33284_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SHOX. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SHOX. Alternate names: GCFX, PHOG, SHOXY, SSApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-014-54943