Learn More
Aviva Systems Biology Corp SHOX2 Antibody - middle region (ARP33286_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33286_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SHOX2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SHOX2. Alternate names: OG12, OG12X, OGI2X, SHOTApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54945