Learn More
Eurofins Discoverx SHP-1, 125 µg. (14-591M)
Supplier:  Eurofins Discoverx 14591M
Encompass
The SHP-1 product is purified N terminal GST-tagged recombinant SHP-1 enzyme. This enzyme is full length. It retains its binding and enzymatic activity and is intended for use in phosphatase assays. This enzyme is provided as 250 μg of 0.494 mg/mL frozen solution. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9016