Learn More
Enzo Life Sciences Shp 1 & 2 substrate (1mg)
SDP
Catalog No. 502011265
Encompass_Preferred
This peptide sequence, EFpYAEVGRSPPDPAK, was selected by phage display analysis for optimal Shp-1 and Shp-2 phosphatase activities. Purity: ≥95% (HPLC). Formulation: Lyophilized. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-1265 Supplier Enzo Life Sciences Supplier No. BMLP1570001
This peptide sequence, EFpYAEVGRSPPDPAK, was selected by phage display analysis for optimal Shp-1 and Shp-2 phosphatase activities. Purity: ≥95% (HPLC). Formulation: Lyophilized. Solubility: Soluble in water. Long Term Storage: -20°C.