Learn More
Eurofins Discoverx SHP-2, 250 µg. (14-622M)
Supplier:  Eurofins Discoverx 14622M
Encompass
The SHP-2 product is purified N terminal GST-tagged recombinant SHP-2 enzyme. This enzyme contains amino acids 230-545. It retains its binding and enzymatic activity and is intended for use in phosphatase assays. This enzyme is provided as 10 μg in frozen solution. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9072