Learn More
Cole-Parmer Cole-Parmer SHP-200D-C-120 Stuart Undergrad Digital Stirring Hot Plate, Ceramic; 120 VAC
Catalog No. NC2181308 Shop All Cole-Parmer Products
Encompass_Preferred
Designed with microprocessor-controlled dual thermocouples, these stirring hot plates offer you accurate temperature control and protection from over-heating. Actual surface temperature is displayed in real time on an easy-to-read digital screen. Powerful magnets and motor provide you with stirring speeds of up to 2000 rpm and mixing capability of up to 15 L of liquid. The square plate design allows you flexibility of vessel sizes and combinations. Up to four 250 mL beakers can easily fit on the surface at the same time. To keep you safe, a hot warning light flashes whenever the plate surface temperature is above 50C and continues to flash until the temperature drops below 50C, even when unplugged. The units also feature BioCote antimicrobial protection which reduces the presence of odor and stain causing microbes such as bacteria, mold, and fungi on the product surface by up to 99.99%.
This item is not returnable. View return policy
Catalog No. NC2181308 Supplier Cole-Parmer Supplier No. 0480763
Designed with microprocessor-controlled dual thermocouples, these stirring hot plates offer you accurate temperature control and protection from over-heating. Actual surface temperature is displayed in real time on an easy-to-read digital screen. Powerful magnets and motor provide you with stirring speeds of up to 2000 rpm and mixing capability of up to 15 L of liquid. The square plate design allows you flexibility of vessel sizes and combinations. Up to four 250 mL beakers can easily fit on the surface at the same time. To keep you safe, a hot warning light flashes whenever the plate surface temperature is above 50C and continues to flash until the temperature drops below 50C, even when unplugged. The units also feature BioCote antimicrobial protection which reduces the presence of odor and stain causing microbes such as bacteria, mold, and fungi on the product surface by up to 99.99%.