Learn More
Selleck Chemical LLC SHP099-5mg
SDP
Catalog No. 502390056
Encompass_Preferred
SHP099 is a highly potent, selective and orally bioavailable small-molecule SHP2 inhibitor with an IC50  value of  71 nM and shows no activity against SHP1.
Catalog No. 50-239-0056 Supplier Selleck Chemical LLC Supplier No. S63885MG
SHP099 is a highly potent, selective and orally bioavailable small-molecule SHP2 inhibitor with an IC50  value of  71 nM and shows no activity against SHP1.