Learn More
Medchemexpress LLC HY-100388 5mg Medchemexpress, SHP099 CAS:1801747-42-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1003885MG
Encompass_Preferred
Medchemexpress, HY-100388 5mg SHP099 CAS:1801747-42-1 SHP099 is a potent, selective, orally available SHP2 inhibitor with an IC 50 of 70 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1805